Order Entry
Finland
ContactUsLinkComponent
Analytical standard, 5-Hydroxyflunixin, reference material, Honeywell Fluka™
Analytical standard, 5-Hydroxyflunixin, reference material, Honeywell Fluka™
Catalog # HONCR1031-10MG
CAS Number:  
Analytical standard, 5-Hydroxyflunixin, reference material, Honeywell Fluka™
Catalog # HONCR1031-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    75369-61-8
  • Description:
    5-Hydroxyflunixin
  • Melting point:
    296…300 °C
  • Formula:
    C₁₄H₁₁F₃N₂O₃
  • Storage temperature:
    Ambient
  • Description:
    5-Hydroxyflunixin

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results