G418 disulphate, Molecular Biology Grade
Catalog # OZBIGS21000
Supplier: OZ BIOSCIENCES
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:692.72 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:108321-42-2
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314