HPLC columns, NUCLEOSIL® N(CH₃)₂
Supplier: MACHEREY-NAGEL
Certificates
About this item
NUCLEOSIL® N(CH₃)₂ is a dimethylamino (DMA) modified silica phases for HPLC.
- Robust spherical silica gel
- Polar and weakly basic anion exchanger
- Separation mechanism based on polar and ionic interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Specifications
- Catalog No:
- MANA721158.30
- MANA720994.46
- Carbon load:
- 4%
- 4%
- pH range:
- 2 - 8
- 2 - 8