Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Giemsa's stain
Giemsa's stain for clinical diagnosis
Giemsa's stain for clinical diagnosis
APPC251338.1610
:
PanReac AppliChem
:
Giemsa's stain for clinical diagnosis
APPC251338.1610
:
PanReac AppliChem
:
Pk:
500 mL
Seq:
2725
Suitability for hematology stain:
passes test
:
:
1230
:
3(6.1),II
:
GHS02
:
GHS05
:
GHS08
:
Ambient
:
51811-82-6
:
MFCD00081642