Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5(6)-TAMRA
5(6)-TAMRA ≥97% (by TLC), Novabiochem®
5(6)-TAMRA ≥97% (by TLC), Novabiochem®
:
Novabiochem (Part of Merck)
5(6)-TAMRA ≥97% (by TLC), Novabiochem®
:
Novabiochem (Part of Merck)
:
98181-63-6
:
MFCD00467499
:
C₂₅H₂₂N₂O₅
:
MFCD16661194
:
430.45 g/mol
:
>300 °C
:
Refrigerator
Identity (IR):
Passes Test
Purity (TLC(011C)):
≥97%
Assay (HPLC, Area%):
≥95.0% (a/a)