Order Entry
Finland
Orders LinkContactUsLinkComponent
Airless dispenser, ecosolution®
Airless dispenser, ecosolution®
Supplier:   SEMADENI
Airless dispenser, ecosolution®
Supplier:  SEMADENI
Certificates
Airless dispenser, ecosolution®
2 Options Available Below
Product Details & Documents
Bottle made of polypropylene.
  • Contracts during dispensing and automatically ensures almost complete emptying of the content
  • Contents are air-protected
  • Dispenser pump is simply snapped on after filling
  • Ideal for viscous products such as lotions or creams
The dispenser pumps have a convenient shutter mechanism (¹/₄ turn to the left). Pumps are available in a short and long spout version (beak). The short version is equipped with an additional, transparent snap cover.
Volume per dosage (stroke): 1,5 ml
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 100 ml
    96 mm
    44 mm
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 236,00 €
    236,00 €
    1 * 100 Items
       236,00 €
  • 200 ml
    131 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 302,00 €
    302,00 €
    1 * 100 Items
       302,00 €