Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Funnels
Filtration Funnels
Filter Funnels
Funnel with grip and air groove
Funnel with grip and air groove
:
RSG
Funnel with grip and air groove
:
RSG
Stainless steel 1.4301.
Very good chemical resistance
Very good temperature resistance
Easy to clean