Order Entry
Finland
ContactUsLinkComponent
Cut resistant gloves, uvex phynomic C5
Cut resistant gloves, uvex phynomic C5
Supplier:  UVEX
Certificates
undefined
Cut resistant gloves, uvex phynomic C5
Supplier:  UVEX
Specifications
  • Catalog No:
  • 112-5372
  • 112-5378
  • 112-5374
  • 112-5377
  • 112-5376
  • 112-5373
  • 112-5375
  • Model:
  • C5
  • C5
  • C5
  • C5
  • C5
  • C5
  • C5
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Dyneema® Diamond Technology/polyamide/elastane
  • Dyneema® Diamond Technology/polyamide/elastane
  • Dyneema® Diamond Technology/polyamide/elastane
  • Dyneema® Diamond Technology/polyamide/elastane
  • Dyneema® Diamond Technology/polyamide/elastane
  • Dyneema® Diamond Technology/polyamide/elastane
  • Dyneema® Diamond Technology/polyamide/elastane
  • Finish:
  • Palm and fingertips coated (Aqua-polymer foam)
  • Palm and fingertips coated (Aqua-polymer foam)
  • Palm and fingertips coated (Aqua-polymer foam)
  • Palm and fingertips coated (Aqua-polymer foam)
  • Palm and fingertips coated (Aqua-polymer foam)
  • Palm and fingertips coated (Aqua-polymer foam)
  • Palm and fingertips coated (Aqua-polymer foam)
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
  • +
  • +
Specifications