Gentamicin sulphate, Ultrapure
Catalog # J16051.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Assay (Unspecified) - Activity Dry Basis:≥590 µg of gentamicin/mg
- Assay (unspecified) Gentamicin Content (C1 + C2b):25 - 50%
- Assay (unspecified) Gentamicin Content (C1a):10 -35%
- Assay (unspecified) Gentamicin Content (C2 + C2a):25 - 55%
- Identity (A, B):Passes Test
- Residue on Ignition:≤1.0%
- Impurity Content Limit of Methanol:≤1.0%
- Loss on Drying:≤18.0%
- Optical Rotation (c=1 in H₂O):+107 to 121°
- pH (4%, H₂O):3.5 - 5.5
Specification Test Results
About this item
:
Merck Index:14,04392
Melting Pt:218…237 °C
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:1405-41-0
MDL Number:MFCD00270181