SPE columns, acrylamide analysis, CHROMABOND® Carbon A
CHROMABOND® Carbon A is a special phase for SPE enrichment of acrylamide from water.
- Activated and highly porous carbon
- Spherical particles
- High specific surface
Application: Enrichment of acrylamide from drinking water according to DIN EN ISO 38413-6.
- Catalog No:
- MANA730167
- MANA730165
- Surface area:
- >1000 m²/g
- >1000 m²/g