ESD safety ankle boots, lace-up, Safety-Grip, Laurenzo Rubber
: ELTEN
Durable ESD safety ankle boots made from robust, breathable cowhide leather, a full padded bellows tongue plus a temperature resistant, dual-density rubber/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
- Climate-regulating, breathable textile lining
- Full length fleece inlay sole (ESD Pro), black
- Soft fleece ESD insole
- Anatomically-shaped insole and ankle support for improved stability
- PU scuff cap protects the leather upper
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in hot or cold areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, HRO, HI
- Catalog No:
- ELTN766941/48
- ELTN766941/42
- ELTN766941/45
- ELTN766941/46
- ELTN766941/47
- ELTN766941/43
- ELTN766941/40
- ELTN766941/44
- ELTN766941/41
- ELTN766941/38
- ELTN766941/39
- ELTN766941/36
- ELTN766941/37
- ELTN766941/49
- ELTN766941/50
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001