ESD safety shoes, lace-up, L10, Shadow
: ELTEN
Casual, vintage style ESD safety shoes for ladies made from cowhide leather, a padded tongue plus a low profile, dual-density TPU/PU sole (L10).
- Leather lining
- Anatomically-shaped insole supports natural heel-to-toe foot movement
- Full length inlay sole (ESD Pro), beige
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey
: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.
- Catalog No:
- 114-8748
- 114-8738
- 114-8739
- 114-8744
- 114-8745
- 114-8746
- 114-8747
- 114-8740
- 114-8741
- 114-8742
- 114-8743
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001