Direct red 80 stain for histology
Catalog # 720-1194
Supplier: RAL
CAS Number:
Specifications
- Pack type:Carton
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:1373.051 g/mol
:1
CAS Number:2610-10-8
Formula:C₄₅H₂₆N₁₀Na₆O₂₁S₆
MDL Number:MFCD00054389