Order Entry
Finland
ContactUsLinkComponent
 
Ammonium acetate ≥98% (by alkalimetry), LiChropur™ for LC-MS, Supelco®
Catalog # 5.33004.0050
Supplier:  Merck
CAS Number:  
undefined
Ammonium acetate ≥98% (by alkalimetry), LiChropur™ for LC-MS, Supelco®
Catalog # 5.33004.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :77.08 g/mol
Melting Pt:114 °C
Density:1.17 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:211-162-9
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066