n-Pentane ≥95%, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6755-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (GC):min. 95 %
- Residue after Evaporation:max. 0.0005 %
- Titrable Acid (meq/g):max. 0.0005
- Water (KF):max. 0.01 %
- at 210 nm:max. 0.4
- at 220 nm:max. 0.15
- at 230 nm:max. 0.05
- at 240 nm:max. 0.01
- at 250 nm:max. 0.004
Specification Test Results
About this item
:
:72.15 g/mol
Boiling Pt:36.1 °C
Density:0.63
Flash Pt:–48 °C
Storage Temperature:Ambient
UN:1265
ADR:3,
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498