3-Acetylthiophene
Catalog # MOLE65251350-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:126.18 g/mol
Boiling Pt:208 °C (1013 hPa)
Melting Pt:56 °C
EINECS:215-996-4
UN:2811
ADR:6.1,III
CAS Number:1468-83-3
Formula:C₆H₆OS
MDL Number:MFCD00005468