Order Entry
Finland
ContactUsLinkComponent
 
 
3-Acetylthiophene 98%
 
undefined
 
3-Acetylthiophene 98%

 

  • Appearance:
    Beige to light brown Adhering crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    57 to 61 °C
  • GC:
    ≥97.5 %