Human Recombinant PSP (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Persephin is a neurotrophic factor of the glial cell line-derived neurotrophic factor (GDNF) family. Persephin promotes survival and growth of dopaminergic and motor neurons, but not peripheral neurons. Persephin is a ligand for the RET receptor tyrosine kinase.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Delivery: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: TT cell proliferation, ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
- Gene ID:O60542
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:PSP
- UniProtKB:O60542
- Protein/peptide name:Persephin
- Purity:≥95%
- Molecular weight:10,4/20,8 kDa
- Sequence:MALSGPCQLW SLTLSVAELG LGYASEEKVI FRYCAGSCPR GARTQHGLAL ARLQGQGRAH GGPCCRPTRY TDVAFLDDRH RWQRLPQLSA AACGCGG
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay