VWR® PerfectBlue Maxi M 'Revolution', Gel system
Catalog # 700-0909
Supplier: VWR Collection
Specifications
- Description:PerfectBlue gel system Maxi M 'Revolution'
- Model:Maxi M 'Revolution'
- Gel volume:250 ml
- Buffer volume:~2000 ml
- Run time:≥60 h
- Gel size W×L:200×250 mm
- Sample capacity:8 – 168
- Unit W×L×H:270×380×155 mm
- Pk:1
Specifications
About this item
Based on the Maxi M model, the Maxi M 'Revolution' offers a built-in buffer circulation system, powered by the system’s electrodes. At the cathode, hydrogen bubbles are produced and pass up a diagonally positioned pipe connected to the two sides of the tank to produce a gentle and continual circulation of the buffer. The system nulls any pH and ionic gradient – without the fuss of mechanical pumps. The system offers 6 comb positions enabling the gel to be partitioned into 2, 3 or 4 areas. In addition, the system can accommodate any of the 23 standard and microtitre comb designs described for the Maxi M model.
*Based on an agarose gel of approximately 5 mm thickness.
Ordering information: Supplied with 3 combs (1,5 mm thickness, 16, 24, 36 teeth), lid, gel tray, 3-point levelling platform, end gates, tank and cables.