
2,4,6-Trichloropyrimidine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3764-01-0
:
Formula:C₄HCl₃N₂
MDL Number:MFCD00006063
:183.42 g/mol
Boiling Pt:213…215 °C
Melting Pt:21…25 °C
Density:1.595
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:223-183-0
ADR:8,III
:GHS05