Order Entry
Finland
ContactUsLinkComponent
2,4,6-Trichloropyrimidine 99%
Irritant
2,4,6-Trichloropyrimidine 99%
Certificates
undefined
Irritant
2,4,6-Trichloropyrimidine 99%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5690 to 1.5710 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information