Order Entry
Finland
ContactUsLinkComponent
Benzyl nicotinate, EMPROVE® api DAB suitable for use as an active pharmaceutical ingredient
Irritant
Benzyl nicotinate, EMPROVE® api DAB suitable for use as an active pharmaceutical ingredient
Catalog # 1.06752.2500
CAS Number:  
Irritant
Benzyl nicotinate, EMPROVE® api DAB suitable for use as an active pharmaceutical ingredient
Catalog # 1.06752.2500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :213.24 g/mol
Boiling Pt:179…180 °C (8 hPa)
Melting Pt:24 °C
Density:1.17 g/cm³ (20 °C)
EINECS:202-332-3
 :GHS07
CAS Number:94-44-0
Formula:C₁₃H₁₁NO₂
MDL Number:MFCD00023584