Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Amino-3-bromo-2-methylpyridine
6-Amino-3-bromo-2-methylpyridine
6-Amino-3-bromo-2-methylpyridine
Irritant
6-Amino-3-bromo-2-methylpyridine
Supplier:
Thermo Scientific
Certificates
Expand image
Irritant
6-Amino-3-bromo-2-methylpyridine
Supplier:
Thermo Scientific
About this item
CAS Number:
42753-71-9
:
Formula:
C₆H₇BrN₂
MDL Number:
MFCD00068230
:
187.04 g/mol
EINECS:
255-927-5
:
GHS07