

2-Bromo-4-methylpyridine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4926-28-7
:
Formula:C₆H₆BrN
MDL Number:MFCD00082590
:172.02 g/mol
Boiling Pt:87 °C (10 mmHg)
Melting Pt:218…228 °C
Density:1.545
Flash Pt:>110 °C (230 °F)
:GHS06
:GHS07
Storage Temperature:Ambient