Nicotinamide
Glass bottle for solids, 1 kg
Catalog # MOLEM66100982
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
- CAS Number:98-92-0
- Formula:C₆H₆N₂O
- MDL Number:MFCD00006395
- Molecular Weight:122.13 g/mol
- Merck Index:12,06574
- Boiling Pt:334 °C (1013 hPa)
- Melting Pt:127…128 °C
- Density:1.4463 g/cm³ (20 °C)
- Flash Pt:182 °C
- Storage Temperature:Ambient
- EINECS:202-713-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



