
2-Chloro-5-iodopyridine 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69045-79-0
:
Formula:C₅H₃ClIN
MDL Number:MFCD01863635
Melting Pt:96…99 °C
UN:2811
ADR:6.1,III
:239.44 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to yellow to light beige Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:96 to 99 °C
- HPLC:≥98.0 %