Order Entry
Finland
Orders LinkContactUsLinkComponent
Pyrene ≥98%

Pyrene ≥98%

Pyrene ≥98%
Certificates
Pyrene ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 129-00-0 Molecular Formula: C16H10 Formula Weight: 202.26 Flash Point: 208°(406°F) Melting Point: 148-152° Boiling Point: 393° Beilstein Reference: 1307225 Merck Reference: 14,7963 MDL No.: MFCD00004136

  • CAS Number:129-00-0
  • Formula:C₁₆H₁₀
  • MDL Number:MFCD00004136
  • Molecular Weight:202.26 g/mol
  • Merck Index:14,7963
  • Boiling Pt:393 °C
  • Melting Pt:148…152 °C
  • Density:1.271
  • Flash Pt:208 °C (406 °F)
  • Storage Temperature:Ambient
  • EINECS:204-927-3
  • UN:3077
  • ADR:9,III
Product Family Options

Product Information