Order Entry
Finland
Orders LinkContactUsLinkComponent
Pyrazine

Pyrazine

Glass bottle for solids, 1 g
Catalog # ROTH5757.1
Marketsource
Supplier:   Roth Carl
Pyrazine
Catalog # ROTH5757.1
Supplier:  Roth Carl
Marketsource
CAS Number:  290-37-9
Pyrazine

Glass bottle for solids, 1 g

CAS Number:  290-37-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:290-37-9
  • Formula:C₄H₄N₂
  • MDL Number:MFCD00006122
  • Molecular Weight:80.09 g/mol
  • Merck Index:13,08047
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:52 °C
  • Density:1.0311 g/cm³ (60.9 °C)
  • EINECS:206-027-6
  • UN:1325
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications