Order Entry
Finland
ContactUsLinkComponent
L(+)-Ascorbic acid 99.0-100.5%, fine powder USP, FCC, J.T.Baker®
L(+)-Ascorbic acid 99.0-100.5%, fine powder USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
L(+)-Ascorbic acid 99.0-100.5%, fine powder USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    99.0 - 100.5%
  • Meets F.C.C. Requirements:
    Passes Test
  • Meets U.S.P Requirements:
    Passes Test
  • Food GMP Manufactured Product:
    20.5 - 21.5°
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.1%
  • Bulk Food Chemical:
    99.0 - 100.5%
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (C₆H₈O₆):
    99.0 - 100.5 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Specific Rotation [Å]²⁵ ^D (+):
    20.5 - 21.5 Degree
  • USP - Residue on Ignition:
    ≤0.1 %
  • FCC - Assay (C₆H₈O₆):
    99.0 - 100.5 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Specific Rotation [Å]²⁵ ^D (+):
    20.5 - 21.5 Degree
  • FCC - Residue on Ignition:
    ≤0.1 %
  • Arsenic (As):
    ≤3 ppm
  • Mesh - Thru U.S. No. 100 Sieve:
    ≥100 %
  • Mesh - Thru U.S. No. 200 Sieve:
    ≥95 %

Specification Test Results

Related Information

Certificates
Search by Lot Number