L(+)-Ascorbic acid 99.0-100.5%, crystalline powder USP, FCC, J.T.Baker®
Catalog # 0936-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Meets F.C.C. Requirements:99.0 - 100.5%
- Food GMP Manufactured Product:Passes Test
- GMP Manufactured Product:Passes Test
- Meets U.S.P Requirements:20.5 - 21.5°
- CAUTION: For Manufacturing, processing or repackaging:≤0.1%
- Bulk Food Chemical:99.0 - 100.5%
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Assay (C₆H₈O₆):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):20.5 - 21.5 Degree
- USP - Residue on Ignition:≤0.1 %
- FCC - Assay (C₆H₈O₆):99.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Specific Rotation [Å]²⁵ ^D (+):20.5 - 21.5 Degree
- FCC - Residue on Ignition:≤0.1 %
- Arsenic (As):≤3 ppm
- Mesh - Thru U.S. No. 20 Sieve:≥100 %
- Mesh - Thru U.S. No. 100 Sieve:≤70 %
- Mesh - Thru U.S. No. 200 Sieve:≤20 %
Specification Test Results
About this item
:
:176.13 g/mol
Merck Index:13,00837
Boiling Pt:553 °C (1013 hPa)
Melting Pt:190…192 °C
Density:1.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328