Hexadecan-16-olide ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 109-29-5 Molecular Formula: C16H30O2 Formula Weight: 254.42 Flash Point: 100°(212°F) Melting Point: 33-34° Beilstein Reference: 148631 MDL No.: MFCD00039668
Synonyms: Oxa-2-cycloheptadecanone, Hexadecanolactone, Oxacycloheptadecan-2-one, 16-Hexadecanolide
- CAS Number:109-29-5
- Formula:C₁₆H₃₀O₂
- MDL Number:MFCD00039668
- Molecular Weight:254.41 g/mol
- Melting Pt:33…34 °C
- Flash Pt:>100 °C (212 °F)
- Storage Temperature:Ambient
- EINECS:203-662-0
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 109-29-5 Molecular Formula: C16H30O2 Formula Weight: 254.42 Flash Point: 100°(212°F) Melting Point: 33-34° Beilstein Reference: 148631 MDL No.: MFCD00039668
Synonyms: Oxa-2-cycloheptadecanone, Hexadecanolactone, Oxacycloheptadecan-2-one, 16-Hexadecanolide
- CAS Number:109-29-5
- Formula:C₁₆H₃₀O₂
- MDL Number:MFCD00039668
- Molecular Weight:254.41 g/mol
- Melting Pt:33…34 °C
- Flash Pt:>100 °C (212 °F)
- Storage Temperature:Ambient
- EINECS:203-662-0
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



