Order Entry
Finland
ContactUsLinkComponent
 
 
 
Titanium(IV) ethanolate 85% tech. contains 5-15% isopropanol
 
undefined
 
 
Titanium(IV) ethanolate 85% tech. contains 5-15% isopropanol

 

  • Appearance:
    Clear colorless to dark yellow Liquid
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥85 %
  • Isopropyl alcohol:
    5 to 15 %