Order Entry
Finland
ContactUsLinkComponent
Titanium(IV) bromide ≥98%
Corrosive
Titanium(IV) bromide ≥98%
Catalog # 39592.09
CAS Number:  
Corrosive
Titanium(IV) bromide ≥98%
Catalog # 39592.09
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :367.5 g/mol
Merck Index:14,9477
Boiling Pt:233 °C
Melting Pt:39 °C
Density:2.6
Storage Temperature:Ambient
EINECS:232-185-0
UN:3260
ADR:8,II
 :GHS05
CAS Number:7789-68-6
Formula:Br₄Ti
MDL Number:MFCD00011265