Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zinc fluoride
Zinc fluoride, anhydrous ≥99%
Zinc fluoride, anhydrous ≥99%
:
Thermo Fisher Scientific
Zinc fluoride, anhydrous ≥99%
:
Thermo Fisher Scientific
:
7783-49-5
:
:
F₂Zn
:
MFCD00011298
:
103.39 g/mol
:
14,10136
:
1500 °C
:
872 °C
:
4.95
:
Ambient
:
232-001-9
:
3288
:
6.1,III
:
GHS05
:
GHS07
msds_ALFA011541
Thermo Scientific Acroseal 25 Years