Order Entry
Finland
ContactUsLinkComponent
 
Zinc chloride 98.0-100.5% (by complexometric analysis), EMPROVE® API USP, Ph. Eur., BP suitable for use as an active pharmaceutical ingredient, SAFC®
Catalog # 1.08815.1000
CAS Number:  
Zinc chloride 98.0-100.5% (by complexometric analysis), EMPROVE® API USP, Ph. Eur., BP suitable for use as an active pharmaceutical ingredient, SAFC®
Catalog # 1.08815.1000
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (Complexometric, ZnCl₂):
    98.0 - 100.5%
  • Identity (Zn):
    Passes Test
  • Identity (Cl):
    Passes Test
  • pH-value (10% water):
    4.6 - 5.5
  • Oxychloride:
    Passes Test
  • Sulfate (SO₄):
    ≤0.01%
  • Total Nitrogen (N):
    ≤0.001%
  • Aluminium, Calcium, Magnesium, Heavy metals, Iron:
    Passes Test
  • As (Arsenic):
    ≤0.0002%
  • Ca (Calcium):
    ≤0.01%
  • Cd (Cadmium):
    ≤0.0005%
  • Fe (Iron):
    ≤0.001%
  • K (Potassium):
    ≤0.15%
  • Na (Sodium):
    ≤0.01%
  • NH₄ (Ammonium):
    ≤0.04%
  • Pb (Lead):
    ≤0.005%
  • Tl (Thallium):
    ≤0.0005%
  • Alkali- and Alkaline Earth Metals (as SO₄):
    ≤1.0%
  • Residual Solvents (Ph. Eur./USP/ICH):
    Excluded by Manufacturing Process

Specification Test Results

About this item

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
 :
 :136.3 g/mol
Merck Index:13,10185
Boiling Pt:732 °C (1013 hPa)
Melting Pt:283 °C
Density:2.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-592-0
UN:2331
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:7646-85-7
Formula:ZnCl₂
MDL Number:MFCD00011295