2-Nitronaphthalene
Glass bottle for solids, 10 mg
Catalog # PROOBCR-307
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:581-89-5
- Formula:C₁₀H₇NO₂
- Molecular Weight:173.17 g/mol
- Boiling Pt:320 °C (1013 hPa)
- Melting Pt:79 °C
- Storage Temperature:Refrigerator
- EINECS:209-474-5
- UN:2538
- ADR:4.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



