Order Entry
Finland
ContactUsLinkComponent
 
Ammonium cerium(IV) sulphate dihydrate ≥94.0%, AnalaR NORMAPUR® analytical reagent
  21232.151
 :  
undefined
Ammonium cerium(IV) sulphate dihydrate ≥94.0%, AnalaR NORMAPUR® analytical reagent
  21232.151
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

 

 

  • Assay:
    Min. 94.0 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Insolubility in diluted H2SO4:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 50 ppm
  • Fe (Iron):
    Max. 50 ppm

 

 

 :
 :632.55 g/mol
 :130 °C
 :231-567-4
 :10378-47-9
 :(NH₄)₂Ce(SO₄)₄·2H₂O
 :MFCD00149417