Ammonium cerium(IV) sulphate dihydrate 95% ACS
:10378-47-9
:
:(NH₄)₂Ce(SO₄)₄·2H₂O
:MFCD00149417
:632.55 g/mol
:130 °C
:231-567-4
:GHS07
:Ambient
- Appearance:Yellow to orange Crystalline powder
- Titration:≥94 %
- Phosphate (PO4):≤0.03 %
- Iron (Fe):≤0.01 %
- Insoluble matter:≤0.05 % (in dilute sulfuric acid)