Order Entry
Finland
ContactUsLinkComponent
6-Methoxy-1-indanone 98%
6-Methoxy-1-indanone 98%
Certificates
undefined
6-Methoxy-1-indanone 98%

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    107 to 111 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information