Order Entry
Finland
ContactUsLinkComponent
 
Dipropyl sulphide for synthesis, Sigma-Aldrich®
  8.41603.0025
 :  Merck
 :  
Dipropyl sulphide for synthesis, Sigma-Aldrich®
  8.41603.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

 

 

  • Melting Point:
    -102 °C
  • Molar Mass:
    118.23 g/mol
  • Boiling Point:
    141 - 142 °C
  • Vapor Pressure:
    8.6 hPa
  • Flash Point:
    28 °C
  • Explosion Limit:
    1 - 51 %(V)
  • Density:
    0.838 g/cm3 (20 °C)

 

 

 :
 :118.24 g/mol
 :12,08053
 :146 °C (1013 hPa)
 :–102 °C
 :0.83 g/cm³ (20 °C)
 :Ambient
 :203-873-8
 :1993
 :3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS07
 :111-47-7
 :C₆H₁₄S
 :MFCD00009379