Dimethyl sulphide 99+%, extra pure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: DMS
- CAS Number:75-18-3
- Formula:C₂H₆S
- MDL Number:MFCD00008562
- Molecular Weight:62.14 g/mol
- Merck Index:12,06204
- Boiling Pt:36.2 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.846 g/cm³
- Flash Pt:–48 °C
- Storage Temperature:Refrigerator
- EINECS:200-846-2
- UN:1164
- ADR:3,II
3 Options Available Below
Product Details & Documents
Synonyms: DMS
- CAS Number:75-18-3
- Formula:C₂H₆S
- MDL Number:MFCD00008562
- Molecular Weight:62.14 g/mol
- Merck Index:12,06204
- Boiling Pt:36.2 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.846 g/cm³
- Flash Pt:–48 °C
- Storage Temperature:Refrigerator
- EINECS:200-846-2
- UN:1164
- ADR:3,II
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4341 to 1.4361 (20°C, 589 nm)
- Water:≤0.3 %
- GC:≥99.0 %



