Order Entry
Finland
ContactUsLinkComponent
Chlorosulphuric acid for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Chlorosulphuric acid for synthesis, Sigma-Aldrich®
Catalog # 8.00220.9050
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Chlorosulphuric acid for synthesis, Sigma-Aldrich®
Catalog # 8.00220.9050
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

About this item

 :
Merck Index:12,02218
Boiling Pt:152 °C (1013 hPa)
Melting Pt:–80 °C
Density:1.75 g/cm³ (20 °C)
ADR:8,I
 :M-Clarity™ quality level = MQ200
 :116.52 g/mol
Storage Temperature:Ambient
EINECS:232-234-6
UN:1754
 :GHS05
 :GHS07
CAS Number:7790-94-5
Formula:HClO₃S
MDL Number:MFCD00011523