Order Entry
Finland
ContactUsLinkComponent
Tetraphenylphosphonium bromide
Tetraphenylphosphonium bromide
Catalog # MOLE11361154-25G
Supplier:  Molekula
CAS Number:  
Tetraphenylphosphonium bromide
Catalog # MOLE11361154-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:299…300 °C
 :419.3 g/mol
EINECS:220-393-4
CAS Number:2751-90-8
Formula:C₂₄H₂₀BrP
MDL Number:MFCD00011915