Choline chloride, EMPROVE® ESSENTIAL DAB 10, FCC, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:67-48-1
:
Formula:HOH₂CCH₂N(CH₃)₃Cl
MDL Number:MFCD00011721
:139.62 g/mol
Merck Index:13,02226
Melting Pt:244…247 °C
Density:∼1.205 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-655-4
Related Information
Frequently Bought Together