0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
(Ethyl)triphenylphosphonium bromide
(Ethyl)triphenylphosphonium bromide ≥98%
(Ethyl)triphenylphosphonium bromide ≥98%
:
Thermo Fisher Scientific
(Ethyl)triphenylphosphonium bromide ≥98%
:
Thermo Fisher Scientific
:
1530-32-1
:
:
C₂₀H₂₀BrP
:
MFCD00011838
:
371.26 g/mol
:
206…210 °C
:
>200 °C (392 °F)
:
Ambient
:
216-223-3
:
3077
:
9,III
:
GHS07
msds_ALFAB23096
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years