Diphosphorus pentoxide
Glass bottle for solids, 1 kg
Catalog # ROTH7609.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride
- CAS Number:1314-56-3
- Formula:P₂O₅
- MDL Number:MFCD00011440
- Molecular Weight:141.94 g/mol
- Merck Index:13,07441
- Melting Pt:562 °C
- Density:2.253 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-236-1
- UN:1807
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



