Dimethyltin oxide
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 2273-45-2 Molecular Formula: (CH3)2SnO Formula Weight: 166.77 Physical Form: Powder Melting Point: dec. UN No.: 3146 MDL No.: MFCD00013838
- CAS Number:2273-45-2
- Formula:C₂H₆OSn
- MDL Number:MFCD00013838
- Molecular Weight:164.78 g/mol
- Melting Pt:Dec.
- Storage Temperature:Ambient
- EINECS:218-881-7
- UN:3146
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
CAS No.: 2273-45-2 Molecular Formula: (CH3)2SnO Formula Weight: 166.77 Physical Form: Powder Melting Point: dec. UN No.: 3146 MDL No.: MFCD00013838
- CAS Number:2273-45-2
- Formula:C₂H₆OSn
- MDL Number:MFCD00013838
- Molecular Weight:164.78 g/mol
- Melting Pt:Dec.
- Storage Temperature:Ambient
- EINECS:218-881-7
- UN:3146
- ADR:6.1,III
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



