

Barium sulphide ≥99.7% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21109-95-5
:
Formula:BaS
MDL Number:MFCD00014192
Merck Index:14,995
Melting Pt:1200 °C
Density:4.25
EINECS:244-214-4
UN:3134
ADR:4.3,III
:169.4 g/mol
:GHS07
:GHS09
Storage Temperature:Ambient