Calcium hydroxide
5 kg
Catalog # SWED12162-5
Supplier: SWED HANDLING
Jump to:
Product Details & Documents
Synonyms: Slaked lime, Calcium dihydroxide
- CAS Number:1305-62-0
- Formula:Ca(OH)₂
- MDL Number:MFCD00010901
- Molecular Weight:74.09 g/mol
- Merck Index:13,01675
- Melting Pt:550 °C
- Density:2.24 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-137-3
- UN:3262
- ADR:8,III
Specifications
Specifications
- Pk:5 kg



