Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Calcium hydroxide ≥95%, TECHNICAL

Plastic bucket, 25 kg
Catalog # 22641.460
Corrosive
Supplier:   VWR Chemicals
Calcium hydroxide ≥95%, TECHNICAL
Catalog # 22641.460
Supplier:  VWR Chemicals
CAS Number:  1305-62-0
Calcium hydroxide ≥95%, TECHNICAL
Calcium hydroxide ≥95%, TECHNICAL

Plastic bucket, 25 kg

CAS Number:  1305-62-0
Price
332,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Slaked lime, Calcium dihydroxide
  • CAS Number:1305-62-0
  • Formula:Ca(OH)₂
  • MDL Number:MFCD00010901
  • Molecular Weight:74.09 g/mol
  • Merck Index:13,01675
  • Melting Pt:550 °C
  • Density:2.24 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-137-3
  • UN:3262
  • ADR:8,III
Specifications

Specification Test Results

  • Assay (at filling):
    Min. 95 %

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg

Specifications